Learn More
Cygnus Technologies Sample Diluent, 1000 mL
Supplier:  Cygnus Technologies I0941000
Encompass
Sample diluent recommended for sample dilutions in the HEK 293 HCP ELISA Kit (F650R), MDCK HCP ELISA Kit (F800), and HeLa HCP ELISA Kit (F810). Offered in three volumes: 100, 500, and 1000 ml.
Catalog No. NC1329723
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.