Learn More
Market Lab Label Tape Dispenser with Blade, 12 rolls
Supplier:  Market Lab 1547
Encompass_Preferred
The Label Tape Dispenser with Blade by MarketLab is a handheld device for dispensing label tape. It features an ergonomic rubber grip handle and a stainless steel cutting blade. The tape holder can accommodate label tape rolls up to 1 inch wide. To use, insert a label tape roll up to 1" wide, pull the desired length of tape, and cut using the built-in blade. It allows for quick and easy dispensing and cutting of label tape. This dispenser helps create custom labels for various uses.
Catalog No. NC2401160
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.