Learn More
LABSYSTEMS DIAGNOSTICS OY Labelled ADA-XALD
Supplier:  LABSYSTEMS DIAGNOSTICS OY 710012002
Encompass_Preferred
NeoMass AAAC 3.0 Internal standard – Labelled ADA-XALD (ISTD – ADA-XALD), is intended to be used for internal quality assurance in clinical laboratories that screen newborns for inborn of metabolism using a tandem mass spectrometer (MS/MS). Labelled ADA-XALD is provided in Lyophilized form.
Catalog No. NC2865473