Learn More
GA International Cryo-DTermo™ – DYMO-Compatible Cryogenic Labels
Supplier:  GA International EC1F076WH
Encompass_Preferred
DYMO-compatible direct thermal labels are ideal for short and long-term cryo storage in liquid or vapor phase liquid nitrogen (-196°C/-321°F), laboratory freezers (-80°C/-112°F), and dry ice transportation. These freezer labels are waterproof and can be immersed in liquid nitrogen immediately after applying. Furthermore, their permanent cryo-adhesive ensures they retain adhesion and can withstand multiple freeze-thaw cycles as well as sterilization by gamma irradiation (up to 50 KGy).
Catalog No. NC1468080