Learn More
Description
- Formulated to meet unique cleaning and materials compatibility requirements found in laboratory glassware washing applications
- High alkaline for effective removal of residues in both soft and hard water
- Contains corrosion inhibitors for excellent materials compatibility
- Reduces utility consumption and cleaning time with low foaming, free-rinsing formulation
- Phosphate-free formulation with biodegradable surfactants
- Manufactured in cGMP compliant, FDA registered and ISO 9001 certified facility
- Specific gravity: 1.15
Specifications
Specifications
| Sterility | Non-sterile |
| Appearance | Yellow |
| Certifications/Compliance | Certificate of analysis |
| Container Type | Plastic jerrican |
| For Use With (Equipment) | Laboratory washers |
| Volume (English) | 5 gal. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.