Learn More
Cytiva LabMate™ PD-10 Buffer Reservoirs
Catalog No. 45000479 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
10 Pack
Catalog No. Quantity
45-000-479 10 Pack
1 options
Catalog No. 45-000-479 Supplier Cytiva Supplier No. 18321603
Add to Cart
Edge
Add to Cart
Use for high-recovery desalting, buffer exchange and removal of low-molecular weight compounds packed with Sephadex™ G-25 Medium. Cytiva Life Sciences™ LabMate™ PD-10 Buffer Reservoirs provide fast, affordable performance.
  • With Disposable PD-10 Desalting Columns, wash and equilibration buffer can be applied in one step with gravity flow protocol
TRUSTED_SUSTAINABILITY
Specifications
Product Type Buffer reservoir
Quantity 10 Pack
Column Type Column Accesory
For Use With (Application) High-recovery desalting and Buffer exchange, as well as removal of low-molecular weight compounds packed with Sephadex G-25 medium, makes easier and more convenient to equilibrate PD-10 column