Learn More
Labnet International, Inc LABNET I5110 MINI INCUBATOR
Supplier:  Labnet International, Inc I5110A
Encompass_Preferred
"Compact Mini Incubator. Labnet's compact Mini Incubator is designed for personal use, small laboratories, and classrooms. The unit features a broad temperature range to meet a variety of small sample applications. Temperature range Ambient +5� to 60�C / Capacity 9.2 liters/0.375 cu. ft. 6 Controls Analog / Exterior dimensions (W x D x H) 11.2 x 11 x 13.2 in (28.5 x 28 x 33.5 cm) / Interior dimensions (W x D x H) 9 x 7.9 x 7.9 in (23 x 20 x 20 cm) / Weight 8.3 kg/18.4 lb / Electrical 120V~ or 230V~, 50/60 Hz"
Catalog No. NC0102962
Only null left
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov