Learn More
Research Products International Corp Simultaneous Start Multi-Channel Timer
SDP
Supplier:  Research Products International Corp 810027
Encompass
This timer is unique in two respects; it is the only multi-channel timer on which various channels can be set independently but started simultaneously. Unit has an hourly chime and also has the loudest alarm we have heard on a timer. Strong enough to be heard above high levels of background noise. Four channels count either up or down to 100 hours with 1-second resolution. A 5th channel works as a calendar and alarm clock. A 6th channel is a 10 hour stopwatch with 1/100 second resolution, lap and split functions. Has a built in stand for benchtop use, clips to a pocket, or sticks to any metal surface with a magnetic back. Powered by two button cell batteries (included).
Catalog No. 50-212-766
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.