Learn More
Grainger INSTOCK Laboratory Utility Deep Ledge, 55PA83
Supplier:  Grainger 55PA83
Encompass_Preferred
The INSTOCK Laboratory Utility Deep Ledge Shelf is a sturdy metal shelf designed for laboratory use. It features a chemical-resistant powder coat finish and measures 36"W x 14"D x 16"H. The shelf has a weight capacity of 200 lbs and can be wall mounted or used freestanding with optional legs. It provides a deep ledge to safely store large containers and equipment. The shelf is made in the USA and offered in gray, white, or black finish.
Catalog No. NC2397825
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.