Learn More
Vernier™ LabQuest 2 and LabQuest Stream Battery
SDPSCI_ED
Catalog No. S16185ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Application):
Educational use only
Catalog No. For Use With (Application)
S16185ND Educational use only
1 options
Catalog No. S16185ND Supplier Vernier™ Supplier No. LQ2BAT
Add to Cart
Add to Cart
Includes
LabQuest 2 and LabQuest Stream Battery
Replacement high-capacity, rechargeable, lithium-ion battery for the Vernier LabQuest 2.
  • This is a replacement high-capacity, rechargeable, lithium-ion battery for the Vernier LabQuest 2, LabQuest Stream, or Go Direct SpectroVis Plus.
  • These batteries include a one-year warranty.
  • With typical school use, however, the batteries will last for three years or more.
  • More information about battery life.
  • If you are replacing a battery, please be sure to dispose of the old battery properly.
Warranty and Services
1 year
Specifications
Type Battery, Replacement Lithium
Warranty 5 Year Limited
For Use With (Application) Educational use only
For Use With (Equipment) LabQuest 2, LabQuest Stream, Go Direct™ SpectroVis™ Plus Spectrophotometer
WARNING: Cancer - www.P65Warnings.ca.gov