Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier LabQuest™ 3
SDPSCI_ED

Catalog No. s1724327 Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Go Direct™ sensors
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
S1724327 Go Direct™ sensors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S1724327 Supplier Vernier Supplier No. LABQ3
Only null left
Add to Cart
Add to Cart

LabQuest™ 3 is a powerful, advanced, easy-to-navigate, and versatile data-logging solution for STEM students.

The LabQuest™ 3 is a standalone data-collection platform that students can use to collect, analyze and interact with data efficiently. With its new capacitive touch screen, students can navigate the platform with ease, and because of its wireless capabilities, students can collect data anywhere with LabQuest™ 3.

  • Connects wirelessly to the entire family of Go Direct™ sensors
  • Easy-to-use platform enables students to generate graphs and analyze results
  • An excellent choice for laboratories, classrooms, or in-the-field investigations

Features

  • Full-featured data-collection platform
  • Compatible with all Vernier sensors
  • Large, high-resolution color screen
  • Touch navigation utilizing gestures
  • Fast data collection with 100,000 samples per second
  • Rechargeable, high-capacity battery
  • Built-in GPS and microphone
  • Wireless connectivity with Wi-Fi and Bluetooth™ wireless technology
  • Built-in apps include Calculator, Periodic Table, Sound Recorder, Stopwatch, and more
  • Student instructions for over 75 of our most popular experiments included

Connectivity to Chromebooks, Computers, iPad™ tablets, and Mobile Devices

Students can share real-time data with multiple devices for a truly hands-on, collaborative learning experience. Use LabQuest 3 to transfer data wirelessly to computers, Chromebooks, or mobile devices running Graphical Analysis™ 4.

USB Sensor Interface

If you want to use your own computer or Chromebook™ to collect data, use LabQuest™ 3 as a conduit between our LabQuest™ sensors and these devices. LabQuest™ 3 works as a USB sensor interface with our Logger Pro™ software or Graphical Analysis 4 app.

Specifications

Type Datalogger
Grade Middle School, High School, College
Interface Wi-Fi 802.11 b/g/n at 2.4 GHz; Bluetooth™ wireless technology
Material Plastic
For Use With (Equipment) Go Direct™ sensors
Temperature Range (Metric) 0°C to 45°C
Weight (English) 1.2 lb.
Electrical Requirements Rechargeable, high-capacity battery; DC charging/powering through external adapter (included)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.