Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Software LabQuest™ Mini
SDPSCI_ED

Catalog No. s16035nd Shop All Vernier Software Products
Change view
Click to view available options
Grade:
High School and College
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S16035ND High School and College
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16035ND Supplier Vernier Software Supplier No. LQMINI
Only null left
Add to Cart
Add to Cart

Includes

Vernier LabQuest Mini interface, Computer connection (USB) cable, User manual, Ordering Information, This product is discontinued and no longer available.

LabQuest Mini is a powerful, affordable, and easy to use sensor interface for computer-based data collection.

  • Use for collecting data with a Windows™ or Macintosh™ computer
  • Works with Logger Pro 3, Logger Lite, or Graphical Analysis for Chrome software
  • Computer connection: USB 2.0 Full Speed
  • Maximum sampling rate: 100,000 samples per second
  • Analog inputs: 3
  • Digital inputs: 2
  • Power is supplied by the USB port of the computer—auxiliary AC power or batteries are not required

Warranty and Services

5-year limited warranty

Specifications

Description LabQuest Mini
Includes Computer connection (USB) cable
Grade High School and College
Interface USB 2.0 Full Speed
For Use With (Application) Sensor interface for computer-based data collection
For Use With (Equipment) Windows XP SP2 or newer, OS X 10.4.11 or newer, Chrome™ OS version 44 or newer
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.