Learn More
Vernier™ LabQuest Viewer
SDPSCI_ED
Catalog No. S16190ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
LabQuest 2 and Original LabQuest
Catalog No. For Use With (Equipment)
S16190ND LabQuest 2 and Original LabQuest
1 options
Catalog No. S16190ND Supplier Vernier™ Supplier No. LQVIEW
Add to Cart
Add to Cart
Includes
LabQuest Viewer
Display and control LabQuest on a computer to teach students how to use LabQuest, demonstrate experiments, or share class data.
  • LabQuest Viewer is computer software that displays and controls the screen of a LabQuest using a computer. When used in conjunction with a projector, this software can be used to demonstrate the functions of LabQuest and to enhance group presentations. This software can also be used to monitor student use of LabQuest devices in your lab, view and control a LabQuest remotely, and easily capture LabQuest screen images for use in lab instructions or assessment questions.
  • View and control one or more LabQuest devices from a computer
  • Use with a projector or interactive white board for class demos and student presentations
  • Display live images of all LabQuests in your lab to monitor student progress throughout the lab
  • Select up to four LabQuest devices for side-by-side comparison of experimental results from different lab groups
  • Capture and share screenshots of the LabQuest screen for use in lab instructions and assessment questions
  • Set customizable permissions that permit viewing without control, or password-protect a connection
  • Install LabQuest Viewer on any computer in your school or college department with the included site license
Warranty and Services
5-year limited warranty
Specifications
Description Connects to LabQuest 2 via Wi-Fi or USB connection
For Use With (Application) Educational use only
For Use With (Equipment) LabQuest 2 and Original LabQuest