Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Labtainer™ 2D BioProcess Container, BioTitan retainers, three ports, Luer locks
GREENER_CHOICE

Catalog No. SH3129403
Change view
Click to view available options
Capacity (Metric):
5 L
10 L
20 L
50 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
SH3129403 20 L
SH3129401 5 L
SH3129402 10 L
SH3129404 50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. SH3129403 Supplier Thermo Scientific™ Supplier No. SH31294.03
Add to Cart
Add to Cart

The Thermo Scientific 2D Labtainer with BioTitan BioProcess Container is composed of Aegis5-14 single-web, multi-layer film.

The Thermo Scientific 2D Labtainer with BioTitan BioProcess Container is composed of Aegis5-14 single-web, multi-layer film.

Features of the Thermo Scientific 2D BPC system include:
• Now manufactured with the BioTitan Retention Device—designed to provide superior universal retention that minimizes the risk of leakage and failure at the connection point
• Use to eliminate post-use cleaning steps required with re-useable containers and to reduce cross-contamination risks
• Constructed in an ISO 7 cleanroom under cGMP conditions

Applications of the Thermo Scientific 2D BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors
• Buffer, culture media, and process liquid preparation and storage
• Chromatography feed and fraction collection
• Harvest, storage, and transport of bulk drug product and bulk drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 20 L
Quantity 20 L
Tubing Style TPE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.