Learn More
Thermo Scientific™ Labtainer™ 2D BioProcess Container, BioTitan retainers, 3 ports, Single Pack

The Thermo Scientific 2D Labtainer™ with BioTitan BioProcess Container is composed of Aegis5-14 single-web, multi-layer film.
$26.75 - $209.00
Specifications
Tubing Length Line 1 | 30 cm (12 in.) |
---|---|
Tubing Length Line 3 | 8 cm (3 in.) |
Tubing Length Line 2 | 30 cm (12 in.) |
Tubing Material Line 3 | Polypropylene |
Tubing Material Line 1 | Polycarbonate |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
SH3129501
![]() |
Thermo Scientific™
SH31295.01 |
5 L |
Each for $209.00
|
|
|||||
SH3129502
![]() |
Thermo Scientific™
SH31295.02 |
10 L |
Each for $28.75
|
|
|||||
SH3129503
![]() |
Thermo Scientific™
SH31295.03 |
20 L |
Each for $26.75
|
|
|||||
Description
The Thermo Scientific 2D Labtainer with BioTitan BioProcess Container is composed of Aegis5-14 single-web, multi-layer film.
Features of the Thermo Scientific 2D BPC system include:
- Now manufactured with the BioTitan Retention Device—designed to provide superior universal retention that minimizes the risk of leakage and failure at the connection point
- Use to eliminate post-use cleaning steps required with re-useable containers and to reduce cross-contamination risks
- Constructed in an ISO 7 cleanroom under cGMP conditions
Applications of the Thermo Scientific 2D BPC include:
- Harvest from, feed into, and collect samples from bioreactors and fermentors
- Buffer, culture media, and process liquid preparation and storage
- Chromatography feed and fraction collection
- Harvest, storage, and transport of bulk drug product and bulk drug precursors
Customize standard BPC systems for optimized single-use technology
Choose from the industry's largest component library with over 20,000 unique components that include:
- Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
- Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
- Filters—used for bioburden reduction; sterilization grade filters and vent filters
- Tubing—thermoplastic elastomers, platinum-cured silicone
- Sample ports—septum-style and needle-free ports
- Clamps—sliders, pinch, and crimp bands
Line 1
MPC insert, polycarbonate
Tubing length: 30 cm (12 in.)
ID x OD: 9.7 x 12.7 mm (0.38 x 0.5 in.)
Line 2
MPC insert, polycarbonate
Tubing length: 30 cm (12 in.)
ID x OD: 9.7 x 12.7 mm (0.38 x 0.5 in.)
Line 3
End plug, polypropylene
Tubing length: 8 cm (3 in.)
ID x OD: 6.35 x 10.92 mm (0.25 x 0.43 in.)
Specifications
30 cm (12 in.) | |
30 cm (12 in.) | |
Polycarbonate | |
TPE |
8 cm (3 in.) | |
Polypropylene | |
Polycarbonate |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.