Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Labtainer™ 2D BioProcess Container, BioTitan retainers, two ports, Luer locks
GREENER_CHOICE

The Thermo Scientific 2D Labtainer with BioTitan BioProcess Container is composed of Aegis5-14 single-web, multi-layer film.

$482.00 - $544.00

Specifications

Tubing Style TPE
View More Specs

Products 6
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
SH3129211
GREENER_CHOICE
View Documents
Thermo Scientific™
SH31292.11
50 mL
Pack of 10 for $516.56
Only null left
Add to Cart
 
SH3129212
GREENER_CHOICE
View Documents
Thermo Scientific™
SH31292.12
100 mL
Pack of 10 for $482.00
Only null left
Add to Cart
 
SH3129213
GREENER_CHOICE
View Documents
Thermo Scientific™
SH31292.13
250 mL
Pack of 10 for $520.41
Only null left
Add to Cart
 
SH3129214
GREENER_CHOICE
View Documents
Thermo Scientific™
SH31292.14
500 mL
Pack of 10 for $544.00
Only null left
Add to Cart
 
SH3129215
GREENER_CHOICE
View Documents
Thermo Scientific™
SH31292.15
1 L
Pack of 10 for $533.72
Only null left
Add to Cart
 
SH3129216
GREENER_CHOICE
View Documents
Thermo Scientific™
SH31292.16
2 L
Pack of 10 for $524.50
Only null left
Add to Cart
 
Description

Description

The Thermo Scientific 2D Labtainer with BioTitan BioProcess Container is composed of Aegis5-14 single-web, multi-layer film.

Features of the Thermo Scientific 2D BPC system include:
• Now manufactured with the BioTitan Retention Device—designed to provide superior universal retention that minimizes the risk of leakage and failure at the connection point
• Use to eliminate post-use cleaning steps required with re-useable containers and to reduce cross-contamination risks
• Constructed in an ISO 7 cleanroom under cGMP conditions

Applications of the Thermo Scientific 2D BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors
• Buffer, culture media, and process liquid preparation and storage
• Chromatography feed and fraction collection
• Harvest, storage, and transport of bulk drug product and bulk drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands

Specifications

Specifications

TPE
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.