Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, Luer Lock connectors, injection site
GREENER_CHOICE

Catalog No. SS00008I
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1 L
2 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
SS00008I 500 mL
SS00006I 100 mL
SS00007I 250 mL
SS00009I 1 L
SS00010I 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. SS00008I Supplier Thermo Scientific™ Supplier No. SS00008I
Add to Cart
Add to Cart

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.

This Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (including an injection site). A wide range of capacities, port configurations and tubing styles are available.

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications:

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Includes 3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site)
Material EVA
Tubing Style EVA
Film Description ASI™ 28
Outer Packaging One 4-mil. poly bag
Port Detail Luer lock connectors, reducer and injection site
Product Line Labtainer
Quantity 50/Cs.
Shipping Condition Room Temperature
Sterilization Method Irradiated
Dimensions (L x W x H) 20.3 x 16.5 cm (8.0 x 6.5 in.)
Length (English) 8 in.
Width (English) 6.5 in.
Product Type Bioprocess Container
Capacity (Metric) 500 mL
Length (Metric) 20.3 cm
Width (Metric) 16.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.