Learn More
Mettler Toledo™ LabX™ Titration Express Starter Pack
Catalog No. 01915034 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
Balances and Titrators
Catalog No. For Use With (Equipment)
01-915-034 Balances and Titrators
1 options
Catalog No. 01-915-034 Supplier Mettler Toledo™ Supplier No. 30097754
Add to Cart
Add to Cart
Connect up to three Mettler Toledo™ instruments at once to optimize workflow, simplify operation, and store results. Includes LabX™ Titration Express, Titrator Instrument License, and an Excellence Balance License for integrated sample weighing.
  • Starter pack includes LabX™ Titration Express, Titrator Instrument License, and an Excellence Balance License for integrated sample weighing
  • Compatible with up to 3 Mettler Toledo instruments at once via an ethernet or USB cable
  • Utilize LabX mail to receive messages or results when away from the titrator or LabX network
  • Excellence Balances connected to LabX have a dedicated entry point for titration tasks that require weighing
  • Schedule tasks to automatically start before coming into the lab, or to complete daily sensor calibrations
Compatibility
  • Titrators: G20, T50, T70, T90, V20, V30, C20, C30
  • Balances: XP, XS, XPE, XSE
TRUSTED_SUSTAINABILITY
Specifications
Type LabX™ Starter Pack (Express Edition)
Display Type No Display
Description Full version of LabX for installation on a single computer
Includes 1 Titrator Instrument License, and 1 Excellence Balance License
For Use With (Equipment) Balances and Titrators