Learn More
Sigma Organic Chemistry LACEY CARBON SUPPORTED GOLD GRID 300 MESH AVERAGE HOLE SIZE 100 MICRON 25/BOX | EA
Supplier:  Sigma Organic Chemistry TEMLC300AU100
Encompass_Preferred
LACEY CARBON SUPPORTED GOLD GRID 300 MESH AVERAGE HOLE SIZE 100 MICRON 25/BOX, EA
About This Item:
UNSPSC Code: 12352200
Storage: ROOM TEMPERATURE
Catalog No. 50-312-5817
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.