Learn More
Sigma Organic Chemistry LACEY CARBON SUPPORTED GOLD GRIDS 400 MESH 50NM HOLE SIZE 5/BOX | LC400AU
Supplier:  Sigma Organic Chemistry TEMLC400AU
Encompass_Preferred
LACEY CARBON SUPPORTED GOLD GRIDS 400 MESH 50NM HOLE SIZE 5/BOX, LC400AU
About This Item:
UNSPSC Code: 12352200
Storage: ROOM TEMPERATURE
Catalog No. 50-312-5902
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.