Learn More
Enzo Life Sciences Lactacystin (1 mg) CAS: 133343-34-7
SDP
Supplier:  Enzo Life Sciences BMLPI1041000
Encompass_Preferred
Key inhibitor for use in proteasome research. A potent and selective irreversible proteasome inhibitor. Specifically inhibits 20S proteasome (MCP; multicatalytic complex). Blocks proteasome activity by targeting the catalytic β-subunit. Has no effect on serine or cysteine proteases. Induces apoptosis inhuman monoblast U937 cells. Inhibits NF-κB activation by inhibiting IκB degradation (IC50=10 µM). Inhibits the ubiquitin proteasome pathway in cell culture. Inhibits cathepsin A. Upregulates HSP70 and HSP22, suggesting an initial neuroprotective pathway. Induces neurite outgrowth in Neuro 2A mouse neuroblastoma cells. Formula: C15H24N2O7S. MW: 376.4. Source: Synthetic.
Catalog No. 50-210-0531
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.