Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactic Acid, 10% v/v, Reagents

Catalog No. 01334847
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-847 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-847 Supplier Reagents Holdings Llc Supplier No. CL100500500A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lactic Acid, 10% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 50-21-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym 2-Hydroxypropanoic Acid, 2-hydroxypropionic Acid
IUPAC Name water
SMILES O

Specifications

pH 1.75
Boiling Point 121°C
Color Colorless to Slight Yellow
Quantity 500 mL
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.022
Percent Purity 10%
Grade Reagent
Chemical Name or Material Lactic Acid, 10% v/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.