Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactic Acid, 10% (v/v), Ricca Chemical
SDP

Catalog No. 427016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
4270-16 500 mL Natural Poly Bottle
4270-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4270-16 Supplier Ricca Chemical Company Supplier No. 427016
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 50-21-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) Mixture
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonym lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
PubChem CID 612
ChEBI CHEBI:78320
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

CAS Min % 89.0, 9.0
CAS Max % 91.0, 11.0
Quantity 500 mL
Packaging Natural Poly Bottle
Percent Purity 10% v/v
Grade Laboratory
Concentration or Composition (by Analyte or Components) 10% (v/v) Aqueous Solution
Chemical Name or Material Lactic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.