Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactic Acid, 85%, FCC, 81-89%, Spectrum™ Chemical
SDP

Catalog No. 18603673 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 mL
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Poly Drum
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-673 500 mL Poly Bottle
18-603-672 125 mL Amber Glass Bottle
18-603-674 6 x 500 mL Poly Bottle
18-603-675 4 L Poly Bottle
18-603-676 4 x 4 L Poly Bottle
18-603-677 20 L Poly Drum
18-603-678 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-603-673 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L1009500MLPL
Add to Cart
Add to Cart

L1009, 50-21-5, C3H6O3

Spectrum™ Chemical Lactic Acid, 85 Percent, FCC is used as a food preservative, curing agent, and flavoring agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 50-21-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

CAS Max % 100%
UN Number UN3265
Percent Purity 81 to 89%
Grade FCC
Quantity 500 mL
Ignition Residue 0.1%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.