Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactic Acid 85%, Purified, Reagents

Catalog No. 01337586
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-586 500 mL
01-337-588 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-586 Supplier Reagents Holdings Llc Supplier No. C2108000500A
Add to Cart
Add to Cart

Lactic Acid 85%, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 50-21-5 , 7732-18-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonym 2-Hydroxypropanoic Acid, 2-hydroxypropionic Acid
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

pH 1.75
Melting Point 16.8°C
Boiling Point 122°C
Color Colorless to Slight Yellow
Grade Pure
Quantity 500 mL
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.206
Chemical Name or Material Lactic Acid 85%, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.