Learn More
Volu Sol Lactophenol Cotton Blue, 4 oz / 120 mL, Volu-Sol™
Supplier: Volu Sol VLC004
Used for mounting medium and a staining agent used in the preparation of slides for microscopic examination of fungi. Fungal elements are stained intensely blue