Learn More
Cayman Chemical LactobIonIc AcId 50g
SDP
Supplier:  Cayman Chemical 1892650 G
Encompass_Preferred
An a2-adrenergic receptor antagonist (Kd 2 6 UM) and an L-type calcium channel blocker (IC50 17 UM) well-known for its coronary vasodilator effects inhibits K-Ras plasma membrane localization (IC50 9 64 UM) blocking the proliferation of K-Ras-transformed tumor cells
Catalog No. 50-298-2348
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.