Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactoferrin Antibody (103), Alexa Fluor™ 488 , R&D Systems™
SDP

Catalog No. NB224838
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB224838 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB224838 Supplier R&D Systems Supplier No. NBP289795AF488

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

Lactoferrin Monoclonal specifically detects Lactoferrin in Human samples. It is validated for Sandwich ELISA Capture.

Specifications

Antigen Lactoferrin
Applications Sandwich ELISA Capture
Classification Monoclonal
Clone 103
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Lactoferrin (P02788; Met1-Lys710).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules
Primary or Secondary Primary
Gene ID (Entrez) 4057
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.