Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lactoferrin Antibody Pair [HRP]
SDP

Catalog No. NB009729 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009729 15 Plates
NB009730 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB009729 Supplier Novus Biologicals™ Supplier No. NBP27934515PLATES
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody Pair

specifically detects in Human samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 78.125 pg/mL
Assay Sensitivity 78.125-5000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Lactoferrin.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin
Gene ID (Entrez) 4057
Gene Symbol LTF
Host Species Rabbit
Kit Contents
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human Lactoferrin monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 0.5 mg/mL of rabbit anti-Human Lactoferrin monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 180 ng of recombinant Human Lactoferrin.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Amino Acids Drugs and other small molecules
    Storage Requirements Storage is content dependent.
    Target Lactoferrin
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 5000 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.