Learn More
Cayman Chemical LactosylceramIdebovIn but 5mg
SDP
Supplier:  Cayman Chemical 271975 MG
Encompass_Preferred
A 5-LO inhibitor (IC50 5 UM for A23187-stimulated 5-LO product synthesis in isolated human granulocytes) inhibits hedgehog-dependent signaling in Shh-LIGHT2 cells and TGF-b- activin A- BMP- or Wnt-dependent signaling in HEK293T cells (IC50s 9 3 3 2 8 2 9 6 and 3 7 UM respectively in luciferase reporter assays) increases TnnT levels in BMP4- and CHIR99021-stimulated human iPSCs
Catalog No. 50-293-8324
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.