Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactulose, MP Biomedicals™

Catalog No. MP215393225 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215393225 25 g
MP215393280 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP215393225 Supplier MP Biomedicals, Inc Supplier No. 0215393225
Add to Cart
Add to Cart

Used in the treatment of chronic constipation, SIBO and hepatic encephalopathy. MP Biomedicals' Lactulose is a synthetic, non-digestible disaccharide (double-sugar) formed from one molecule each of fructose and galactose (monosaccharides). It is also used

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4618-18-2
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.297
InChI Key JCQLYHFGKNRPGE-DJYZFUSFSA-N
Synonym lactulose
PubChem CID 131839611
IUPAC Name (2S,3S,4S,5R,6R)-2-[(2R,4S,5R)-4,5-dihydroxy-2,5-bis(hydroxymethyl)oxolan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C2O)(CO)O)CO)O)O)O)O

Specifications

pH 5.5 to 6.5 (1% aq. soln.)
Quantity 25 g
Solubility Information Soluble in 5% of water (colorless)
Optical Rotation −45 to −52° (c=5, water, 24 hrs)
Formula Weight 342.3
Percent Purity ∼98%
Physical Form Powder
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.