Learn More
Novus Biologicals pI Standard Ladder 1, 175 uL
Supplier: Novus Biologicals 040644
pI Standard Ladder 1: 4.0, 4.9, 6.0, 6.4, 7.3 (175 uL); Ships overnight at room temperature. Upon receipt, store at 4°C.