Learn More
BURKLE INC Liquid funnel sterile
Supplier: BURKLE INC 53786002
Disposable funnel for reliable filling of liquids ideal for filling bottles and other containers with a narrow filler opening The disposable funnel ensures no loss when transferring liquid samples or free-flowing solids That makes this practical and hygienic ping aid a helpful accessory for sampling or in laboratories Clean room produced funnel with suspension eye and venting fins for ventilation to ensure uniform flow while ping In combination with a filter paper the disposable funnel is also well suited for filtering
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.