Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Laemmli SDS Sample Buffer with Pyronin Y, reducing, 4X

Catalog No. AAJ62115AD
Change view
Click to view available options
Quantity:
25 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62115AD 50 mL
AAJ62115AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62115AD Supplier Thermo Scientific™ Supplier No. J62115AD
Add to Cart

missing translation for 'validMainframeMsg'

For protein sample preparation to be used in the Laemmli SDS-PAGE system

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Color Pink
Concentration 4X
Form Liquid
For Use With (Application) Laemmli SDS-PAGE
Quantity 50 mL
Product Type Sample Loading Buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.