Learn More
Bioworld Laemmli SDS-Sample Buffer (6X), Non-Reducing, 50 mL
Supplier:  Bioworld 105700192
Encompass_Preferred
Laemmli (SDS-Sample) 6X Buffer, Non-Reduced An electrophoretic dye for denaturation of proteins and monitoring the front of running gel. Composition of this buffer is similar to the reducing buffer minus mercaptoethanol. Technical Information:Ultra Pure GradeContents of a 1X Solution: 2% SDS and 10% glycerol, 62.5mM Tris -HCl (pH 6.8), 0.0025% Bromophenol blue.DNase: None detectedRNase: None detectedProteases: None detectedStability (4°C): for 1year
Catalog No. 50-196-781
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.