Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAG-3 Antibody Pack, R&D Systems™
SDP

Catalog No. NB397896 Shop All Novus Products
Change view
Click to view available options
Quantity:
3 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397896 3 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397896 Supplier R&D Systems Supplier No. NBP311743

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. nan Monoclonal Antibody

LAG-3 Monoclonal antibody specifically detects LAG-3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry

Specifications

Antigen LAG-3
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation See individual datasheets
Gene Alias 17b4, 17b4 lag3, 17b4 neutralizing, anti human lag3, Anti-LAG-3, CD223, CD223 Antigen, Human LAG3, Human LAG-3, LAG3, LAG-3, LAG-3 17b4, LAG-3 blocking, lymphocyte activating 3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Secreted lymphocyte activation gene 3 protein, sLAG-3
Immunogen See individual datasheets
Quantity 3 Vials
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Immunology, Innate Immunity
Gene ID (Entrez) 3902
Target Species Human
Content And Storage Storage is content dependent.
Form Purified
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.