Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAG3 Antibody, ProSci
SDP

Catalog No. 16595231 Shop All ProSci Products
Change view
Click to view available options
Quantity:
0.1 mg
20 μg Trial Size
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-231 20 μg Trial Size
16-919-523 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-595-231 Supplier ProSci LLC Supplier No. 86570020
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Specifications

Antigen LAG3
Applications ELISA, Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution LAG3 antibody can be used for Western blot at 1 μg/mL. Antibody can also be used for Immunohistochemistry at 2 μg/mL. For Immunoflorescence start at 20 μg/mL. Antibody validated: Western Blot in human samples; Immunohistochemistry in human samples and Immunofluorescence in human samples. All other applications and species not yet tested.
Formulation LAG3 Antibody is supplied in PBS containing 0.02% sodium azide.
Gene Accession No. NP_002277
Gene Alias LAG3 Antibody: lymphocyte activating 3, LAG-3, CD223
Host Species Rabbit
Purification Method LAG3 Antibody is affinity chromatography purified via peptide column.
Quantity 20 μg Trial Size
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3902
Target Species Human
Content And Storage LAG3 antibody can be stored at 4°C for three months and -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Product Type Antibody
Form Liquid
Isotype IgG
Research Category Immunology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.