Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUJIFILM Wako LAL Reagent Water

Catalog No. p-7217324
Encompass
Change view
Click to view available options
Fill Volume:
30 mL
125 mL
Includes:
12 x 125 mL plastic bottles
20 x 30 mL glass bottles
Quantity:
12 Bottles
20 Bottles
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fill Volume Includes Quantity
12-333-035 30 mL 20 x 30 mL glass bottles 20 Bottles
12-333-036 125 mL 12 x 125 mL plastic bottles 12 Bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-333-035 Supplier FUJIFILM Wako Supplier No. LRW2030
Only null left
Add to Cart
Add to Cart

Endotoxin-free water for use in all aspects of endotoxin testing

  • Endotoxin-free water (<0.001 EU/mL)
  • Steam-sterilized by USP standards
  • Non-LAL reactive
  • Great for reconstitution of LAL, RSE and CSE
  • Used to perform dilution series of RSE and CSE
  • Used for sample dilutions

Specifications

Form Liquid
Fill Volume 30 mL
Includes 20 x 30 mL glass bottles
Quantity 20 Bottles
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.