Learn More
Description
Specifications
Specifications
| Antigen | Lambda Light Chain |
| Applications | ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | RM127 |
| Concentration | 1 mg/mL |
| Conjugate | Biotin |
| Dilution | Flow Cytometry 0.2ug/ml-2ug/ml, ELISA 0.05ug/ml - 0.2ug/ml, Immunohistochemistry 0.5ug/ml - 2ug/ml, Immunocytochemistry/Immunofluorescence 0.5ug/ml - 2ug/ml, Immunocytochemistry 0.5 - 2 ug/mL |
| Formulation | PBS, 50% Glycerol, 1% BSA with 0.09% Sodium Azide |
| Gene Alias | IGLC, immunoglobulin lambda constant group |
| Gene Symbols | IGLC1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
