Learn More
Santa Cruz Biotechnology LAMIN A/C (636)
Supplier:  Santa Cruz Biotechnology SC7292
Encompass
Lamin A/C (636),mouse monoclonal IgG2b; 200 ug/ml,recommended for detection of Lamin A and Lamin C of human and porcine origin by WB, IP, IHC(P) and FCM,agarose conjugate for IP studies,500 ug/0.25mL agarose
Catalog No. NC9749697
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.