Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin A/C, Mouse anti-Monkey, Syrian Hamster, Rat, Mouse, Human, Clone: 4C11, Millipore Sigma™

Catalog No. MABT134125 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT134125 25 μg
MABT1341100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABT134125 Supplier MilliporeSigma Supplier No. MABT134125UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Lamin A/C, clone 4C11, Cat. No. MABT1341, is a highly specific mouse monoclonal antibody that targets Lamin A/C and has been tested for use in Immunofluorescence, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP), and Western Blotting.

Prelamin-A/C (UniProt: P02545) is encoded by the LMNA (also known as LMN1) gene (Gene ID: 4000) in human. It is cleaved into Lamin-A/C (also known as 70 kDa Lamin, Renal carcinoma antigen NY-REN-32). Lamins are components of the nuclear lamina that provide a framework for the nuclear envelope and may also interact with chromatin. Lamin A and C are present in equal amounts in the lamina of mammals. Plays an important role in nuclear assembly, chromatin organization, nuclear membrane and telomere dynamics. Lamin A is initially synthesized as prelamin A that undergoes several modifications in the carboxyl terminal region that allow incorporation of prelamin A into the nuclear envelope and its subsequent processing into the mature lamin A. Cleavage of 15 residues (aa 647-662) by ZMPSTE24/FACE1 generates the final protein product. Unlike mature lamin A, prelamin A accumulates as discrete and localized foci at the nuclear periphery. Prelamin-A/C can accelerate smooth muscle cell senescence. It can act to disrupt mitosis and induce DNA damage in vascular smooth muscle cells (VSMCs), leading to mitotic failure, genomic instability, and premature senescence. Mutations in LMNA gene are known to cause Emery-Dreifuss muscular dystrophy that is characterized by weakness and atrophy of muscle without involvement of the nervous system. Some mutations have also been linked to familial type of lipodystrophy characterized by the loss of subcutaneous adipose tissue in the lower parts of the body. (Ref.: Casasola, A., et al. (2016). Nucleus 7(1); 84-102).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lamin A/C
Applications ChIP Assay, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4C11
Conjugate Unconjugated
Dilution Western Blotting Analysis: A representative lot detected Lamin A/C in Western Blotting applications (Gesson, K., et. al. (2016). Genome Res. 26(4):462-73; Roblek, M., et. al. (2010). PLoS One. 5(5):e10604).Dot Blot Analysis: A representative lot detected Lamin A/C in Dot Blot applications (Roblek, M., et. al. (2010). PLoS One. 5(5):e10604). Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Lamin A/C in HeLa cells.Western Blotting Analysis: A representative lot detected Lamin A/C in WB used to demonstrate high titration and excellent properties of clone 4C11 (Courtesy of Marie lang, M.D., Stefan Schuchner, Ph.D. and Egon Ogris, M.D., Medical University of Vienna, Austria).Immunofluorescence Analysis: A representative lot detected the Lamin A/C species at the nuclear lamina of HeLa cells (Courtesy of Marie Lang, M.D., Stefan Schuchner, Ph.D. and Egon Ogris, M.D., Medical University of Vienna, Austria).Immunoprecipitation Analysis: A representative lot detected Lamin A/C in WB used to demonstrate high titration and excellent properties of clone 4C11 (Courtesy of Marie Lang, M.D., Stefan Schuchner, Ph.D. and Egon Ogris, M.D., Medical University of Vienna, Austria).Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected Lamin A/C in Chromatin Immunoprecipitation applications (Gesson, K., et. al. (2016). Genome Res. 26(4):462-73).Immunofluorescence Analysis: A representative lot detected Lamin A/C in Immunofluorescence applications (Roblek, M., et. al. (2010). PLoS One. 5(5):e10604).Immunoprecipitation Analysis: A representative lot immunoprecipitated Lamin A/C in Immunoprecipitation applications (Gesson, K., et. al. (2016). Genome Res. 26(4):462-73).
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias 70 kDa Lamin;Renal carcinoma antigen NY-REN-32
Gene Symbols LMNA;LMN1
Host Species Mouse
Immunogen Purified protein corresponding to the Ig-fold of human Lamin A/C.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001244303
Test Specificity Clone 4C11 detects Lamin A/C at the nuclear lamina in multiple species.
Target Species Monkey, Syrian Hamster, Rat, Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.