Learn More
Invitrogen™ Lamin A (Cleaved Asp230) Polyclonal Antibody

Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with full length lamin A or lamin C.
Specifications
Specifications
| Antigen | Lamin A (Cleaved Asp230) |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 |
| Gene | LMNA |
| Gene Accession No. | P02545, P48678, P48679 |
| Gene Alias | 70 kDa lamin; CDCD1; CDDC; CMD1A; CMT2B1; Dhe; EMD2; FPL; FPLD; FPLD2; HGPS; IDC; lamin; lamin A; lamin A/C; lamin A/C-like 1; lamin C; lamin C2; lamin-A/C; LDP1; LFP; LGMD1B; Lmn1; LMNA; LMNC; LMNL1; OTTHUMP00000015843; OTTHUMP00000015846; OTTHUMP00000015847; Prelamin-A/C; PRO1; Renal carcinoma antigen NY-REN-32 |
| Gene Symbols | LMNA |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.