Learn More
Essendant LAMINATORHALO 125WH
Supplier:  Essendant FEL5753101
Encompass_Preferred
User-friendly home office personal laminator InstaHeat Technology delivers no wait rapid one-minute warm up Offers versatile hot lamination of 3 mil or 5 mil pouches as well as cold pouches The convenient release lever allows easy pouch re-centering or removal Auto Shut-off saves energy and prevents overheating Laminator Supply Type- Hot/Cold Use Rating- Moderate Electric/Manual/Battery- Electric Number of Rollers- 2
Catalog No. 50-000-35107
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.