Learn More
CPC Scientific H-Cys-Asp-Pro-Gly-Tyr-Ile-Gly-Ser-Arg-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific LAMN004B
Encompass
Peptide derived from residues 925-933 of the laminin B1 chain that binds to the laminin receptor but has lower affinity compared with the pepetide amide of the same sequence.SEQUENCE: H-Cys-Asp-Pro-Gly-Tyr-Ile-Gly-Ser-Arg-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: CDPGYIGSRMOLECULAR FORMULA: C40H62N12O14S1MOLECULAR WEIGHT: 967.1STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [110590-60-8]SYNONYMS: Laminin -1 Chain (925-933) (human, mouse)RESEARCH AREA: CancerREFERENCES:Y. Iwamoto, et al., Science 238, 1132 (1987)SKU(s): LAMN-004A, LAMN-004B
Catalog No. 50-210-9630
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.