Learn More
Supply Solutions Laminin, 114956-81-9, MFCD00081739, 1mg (2mL)
Supplier:  Supply Solutions SIGMA 11243217001
Encompass_Preferred
Laminin is purified as laminin-nidogen complex from mouse Engelbreth-Holm-Swarm (EHS) sarcoma. Laminin is a heterotrimer made from ?, ? and ? chain subunits. It is a glycoprotein which is required for the structural scaffolding of basement membranes. Laminin supports growth and differentiation of many cell types including epithelial, endothelial, neural, muscle and liver cells.
Catalog No. 50-100-3381
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.