Learn More
Advanced Biomatrix iMatrix-111 Recombinant Laminin, 0.35 mg
Supplier:  Advanced Biomatrix 54062EA
Encompass_Preferred
Laminin-111 protein plays an important role in embryonic development. iMatrix-111 is unique in that no pre-coating steps are necessary. This kit comes with 2 vials for a total of 0.35 mg of protein. Concentration: 0.5 mg/mL. Laminin 111 supports the survival, proliferation, and differentiation of many different cell types in vitro. However, its distribution after birth is restricted to only a few tissues, such as the brain and kidney. During development, Riechert’s membrane supporting the extraembryonic trophoblasts express laminin 111. Reichert’s membrane is essential for epiblast differentiation, which is essential for the formation of the three embryonic germ layers by pluripotent cells in the inner cell mass. In the adult, laminin α1 chain expression is relatively restricted and is only present in some epithelial basement membranes, and primarily found in tissues such as the eye, liver, and kidney, whereas hardly expressed at all in endothelial, adipose
Catalog No. NC2368628
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.