Learn More
Advanced Biomatrix iMatrix-221, Recombinant Laminin 221, 0.5 mg/ml, 6 x 0.35 mL
Supplier:  Advanced Biomatrix 53466EA
Encompass_Preferred
Laminin-221 is found predominantly in the basal lamina of striated muscle tissues. Expression deficiencies are implicated in muscular dystrophy and cardiomyopathies. iMatrix-221 is unique in that no pre-coating steps are necessary. This kit comes with 6 vials for a total of 1.05 mg of protein.
Catalog No. NC2410845
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.