Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Laminin (925-933)

Catalog No. AAJ64340MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64340MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64340MCR Supplier Thermo Scientific Chemicals Supplier No. J64340MCR
Add to Cart

For cellular and molecular biology applications

Derived from residues 925-933 of human laminin B1, this peptide binds the laminin receptor, Inhibits experimental metastasis. find useful for product applications. And it is also used in Western Blot, Blocking Peptides, Dot Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Flow Cytomety, Cell Culture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
For Use With (Application) Derived from residues 925-933 of human laminin B1, this peptide binds the laminin receptor
Molecular Weight (g/mol) 967.05
Product Type Laminin (925-933)
Quantity 1 mg
Regulatory Status RUO - research use only
Sequence Cys-Asp-Pro-Gly-Tyr-Ile-Gly-Ser-Arg

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.