Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Laminin alpha-3 Recombinant Rabbit Monoclonal Antibody (JJ0957)
GREENER_CHOICE

Catalog No. PIMA541201 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541201 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA541201 Supplier Invitrogen™ Supplier No. MA541201
  • $606.00
    $504.65 / Each of 1

    Save $101.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

Laminin alpha-3 Recombinant Monoclonal Antibody for ICC/IF, Western Blot

Laminin is a non-collagenous protein of basement membranes. By rotary shadowing it has a cross-like shape, consisting of one long arm, (400 kD) and three short arms (200 kD). Antibodies to laminin localize exclusively in basement membranes. Laminin is found throughout the basement membrane, but the reaction with the antibodies to laminin is more intense in the lamina lucida, the part of the basement membrane adjacent to the cell membrane. Laminin binds to type IV collagen, a component exclusively localized in the basement membrane. Laminin has been shown to play a role in cell adhesion and attachment in vivo and in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin alpha-3
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JJ0957
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene LAMA3
Gene Accession No. Q16787
Gene Alias [a]3B; BM600; BM600 150kD subunit; E170; Epiligrin; Epiligrin 170 kDa subunit; epiligrin alpha 3 subunit; epiligrin subunit alpha; kalinin 165kD subunit; kalinin subunit alpha; LAM3, alpha-3 subunit; Lama3; Lama3B; laminin 5 alpha 3; laminin 5, alpha-3 subunit; laminin A3; laminin subunit alpha 3; laminin subunit alpha-3; laminin, alpha 3; laminin, alpha 3 (nicein (150kD), kalinin (165kD), BM600 (150kD), epilegrin); Laminin-5; laminin-5 alpha 3 chain; laminin-5 subunit alpha; Laminin-6 subunit alpha; laminin-7 subunit alpha; LAMNA; LOCS; nicein 150kD subunit; nicein subunit alpha; nicein, 150kDa
Gene Symbols LAMA3
Host Species Rabbit
Immunogen Recombinant protein within human Laminnin 5 alpha 3 aa 50-150.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3909
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.