Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin alpha 4 Antibody (CL3185) Alexa Fluor™ Plus 488, R&D Systems™
SDP

Catalog No. RD10130064
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10130064 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10130064 Supplier R&D Systems Supplier No. NBP344409AFP488

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Laminin alpha 4 Monoclonal antibody specifically detects Laminin alpha 4 in Human,Mouse samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)

Specifications

Antigen Laminin alpha 4
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL3185
Conjugate Alexa Fluor Plus 488
Formulation 50mM Sodium Borate
Gene Alias DKFZp686D23145, LAMA3LAMA4*-1, laminin alpha 4 chain, laminin subunit alpha-4, laminin, alpha 4, Laminin-14 subunit alpha, Laminin-8 subunit alpha, Laminin-9 subunit alpha
Host Species Mouse
Immunogen This antibody was generated using a recombinant protein sequence of Q16363, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3910
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.